Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Tributylstannyl)thiophene

Supplier: APOLLO SCIENTIFIC
Danger

OR25730-100G OR25730-10G OR25730-50G
76611-420EA 177 USD InStock
76611-420 76611-422 76611-424
2-(Tributylstannyl)thiophene
2-(Tributylstannyl)thiophene


Formula: C₁₆H₃₀SSn
MW: 373.16
Boiling Pt: 155 °C/0.1mmHg
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00192513
CAS Number: 54663-78-4
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty