Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-1,3-propanediamine

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: N-Boc-1,3-propanediaminetert-Butyl (3-aminopropyl)carbamatetert-Butyl N-(3-Aminopropyl)carbamateN-Boc-1,3-diaminopropaneN-(3-Aminopropyl)carbamic Acid tert-Butyl EsterN-1-Boc-1,3-diaminopropane·HClN-(tert-Butoxycarbonyl)-1,3-propanediamine

OR1269-5G OR1269-25G OR1269-1G
76615-586EA 38 USD InStock
76615-586 76615-588 76615-584
N-Boc-1,3-propanediamine
N-Boc-1,3-propanediamine


Formula: C₈H₁₈N₂O₂
MW: 174.244 g/mol
Boiling Pt: 203 °C
Melting Pt: 22 °C
Density: 0.998 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00210021
CAS Number: 75178-96-0
UN: 3259
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty