Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-1,4-Diazabicyclo[4.3.0]nonane, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR15658-5G OR15658-1G
76624-830EA 86 USD InStock
76624-830 76624-832
(R)-1,4-Diazabicyclo[4.3.0]nonane, Technical Grade
(R)-1,4-Diazabicyclo[4.3.0]nonane


Formula: C₇H₁₄N₂
MW: 126.20 g/mol
Boiling Pt: 65…69 °C/3mmHg
Storage Temperature: Ambient
MDL Number: MFCD09752820
CAS Number: 96193-27-0
UN: 3267
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty