Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dess-Martin periodinane

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: Acetic acid 1,1-diacetoxy-3-oxo-1l5-ioda-2-oxa-indan-1-yl esterDMP1,1-Dihydro-1,1,1-triacetoxy-1,2-benziodoxol-3(1H)-one(1,1,1-Triacetoxy)-1,1-dihydro-1,2-benziodoxol-3(1H)-one

OR14263-25G OR14263-100G OR14263-250G
76611-130EA 177 USD InStock
76611-130 76611-132 76611-128
Dess-Martin periodinane
Dess-Martin periodinane


Formula: C₁₃H₁₃IO₈
MW: 424.15 g/mol
Melting Pt: 130…133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00130127
CAS Number: 87413-09-0
UN: 1479
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty