Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-methoxypropionic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR15940-5G OR15940-1G
76620-694EA 213 USD InStock
76620-694 76620-692
2-Chloro-3-methoxypropionic acid
2-Chloro-3-methoxypropionic acid


Formula: C₄H₇ClO₃
MW: 138.55 g/mol
Boiling Pt: 155 °C/24.8mmHg
Density: 1.3 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03093624
CAS Number: 2544-05-0
UN: 3265
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty