Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(+)-Glucosamine hydrochloride 98+%

Supplier: APOLLO SCIENTIFIC
Warning
Synonyms: (2R,3R,4S,5R)-2-Amino-3,4,5,6-tetrahydroxyhexanal hydrochlorideD-Glucosamine hydrochloride2-Amino-2-deoxy-D-glucopyranose Hydrochloride

OR5282-250G OR5282-5KG OR5282-1KG
76591-374EA 16 USD InStock
76591-374 76591-376 76591-378
D-(+)-Glucosamine hydrochloride 98+%
D-(+)-Glucosamine hydrochloride


Formula: C₆H₁₃NO₅
MW: 215.63 g/mol
Melting Pt: ca 192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00135831
CAS Number: 66-84-2
EINECS: 200-638-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty