Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

indane-5-sulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger
Synonyms: 2,3-Dihydro-1H-indene-5-sulfonyl chloride2,3-Dihydro-1H-indene-5-sulphonyl chloride

OR3684-1G OR3684-5G
76621-110EA 58 USD InStock
76621-110 76621-108
indane-5-sulfonyl chloride
indane-5-sulfonyl chloride


Formula: C₉H₉ClO₂S
MW: 216.679 g/mol
Melting Pt: 47.8…48.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05237217
CAS Number: 52205-85-3
UN: 3261
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty