Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-4-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

12167-0250 12167-1000
TS12167-1000EA 184 USD InStock
TS12167-1000 TS12167-0250
3-Hydroxy-4-nitrobenzoic acid 98%
3-Hydroxy-4-nitrobenzoic acid


Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 233 °C
Storage Temperature: Ambient
MDL Number: MFCD00007110
CAS Number: 619-14-7
EINECS: 210-580-9
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point ≥228 °C
Water ≤0.5 %
Titration with NaOH ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty