Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™

Supplier: Thermo Scientific Chemicals
Synonyms: Perfluorotri-n-butylaminePerfluorotributylamineTris(nonafluorobutyl)aminePFTBAFC-43HeptacosafluorotributylamineHeptacosafluorotri-n-butylamine

12377-2500 12377-0100 12377-1000
TS12377-0100EA 96 USD InStock
TS12377-0100 TS12377-1000 TS12377-2500
Tris(perfluorobutyl)amine , Perfluoro-compound FC-43™
Tris(perfluorobutyl)amine


Formula: C₁₂F₂₇N
MW: 671.1 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –52 °C
Density: 1.88 g/cm³
MDL Number: MFCD00000436
CAS Number: 311-89-7
EINECS: 206-223-1
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Boiling point 173 to 181 °C
Density (25 °C) 1.865 to 1.895 g/ml
Additional info distillation range (between 165 and 185 °C): > 97%
Additional info residue: < 10 ug/ml
Additional info pour point: < - 40 °C
Additional info dielectric strength: > 35 kV
Viscosity 2.3 to 3.3 cSt (at 25 °C)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty