Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-Mandelic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (R)-(-)-2-Hydroxy-2-phenylacetic acid(R)-2-Hydroxy-2-phenylacetic acidD(-)-2-Hydroxy-2-phenylacetic acidD(-)-Mandelic acid

12532-0250 12532-1000 12532-0050
TS12532-1000EA 161 USD InStock
TS12532-1000 TS12532-0250 TS12532-0050
(R)-(-)-Mandelic acid 99%
(R)-(-)-Mandelic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 130…135 °C
Flash Pt: >190 °C
Storage Temperature: Ambient
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6
Order Now
Specification Test Results
Appearance White to light yellow to light beige Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 130 to 135 °C
Water ≤1 % (K.F.)
Sulfated ash ≤0.2 %
Specific optical rotation -152° to -158° (20 °C, 589 nm) (c=2, H2O)
Titration with NaOH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty