Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oxamic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-2-oxoacetic acid

12962-0100 12962-0250 12962-1000
TS12962-1000EA 276 USD InStock
TS12962-1000 TS12962-0250 TS12962-0100
Oxamic acid 98%
Oxamic acid


Formula: C₂H₃NO₃
MW: 89.05 g/mol
Melting Pt: 210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00008006
CAS Number: 471-47-6
EINECS: 207-443-0
Order Now
Specification Test Results
Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Titration RCOCl ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty