Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Purine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 1H-Purine

13167-0050 13167-0010
TS13167-0050EA 761 USD InStock
TS13167-0050 TS13167-0010
Purine 99%
Purine


Formula: C₅H₄N₄
MW: 120.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00079221
CAS Number: 120-73-0
EINECS: 204-421-2
Order Now
Specification Test Results
Appearance White to light yellow to beige Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with HClO4 ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty