Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyrrole-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

13207-0050 13207-0010 13207-0250
TS13207-0250EA 402 USD InStock
TS13207-0250 TS13207-0050 TS13207-0010
Pyrrole-2-carboxylic acid 97%
Pyrrole-2-carboxylic acid


Formula: C₅H₅NO₂
MW: 111.10 g/mol
Melting Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00005219
CAS Number: 634-97-9
Order Now
Specification Test Results
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty