Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Tartaric acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: unnatural tartaric acid(2S,3S)-2,3-Dihydroxysuccinic acid(S,S)-(-)-2,3-Dihydroxysuccinic acid(S,S)-(-)-Tartaric acid

13786-1000 13786-2500 13786-0010 13786-0250
TS13786-0250EA 46.51 USD InStock
TS13786-0250 TS13786-2500 TS13786-0010 TS13786-1000
D-(-)-Tartaric acid 99%
D(-)-Tartaric acid


Formula: C₄H₆O₆
MW: 150.09 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 166…170 °C
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6
Order Now
Specification Test Results
Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 167 to 171 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 3 hrs)
Sulfated ash ≤0.1 %
Specific optical rotation -11.0° to -13.0° (20 °C, 589 nm) (c=20, water)
Titration with NaOH ≥98.5 %
Heavy metals (as Pb) ≤10 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty