Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Malic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: L-(–)-Malic Acid(S)-(-)-Hydroxysuccinic acidL(-)-Hydroxy-1,4-butanedioic acid(S)-(-)-Hydroxy-1,4-butanedioic acid(S)-2-hydroxysuccinic acidL(-)-Hydroxysuccinic acidL(-)-Malic acid

15059-0250 15059-5000 15059-1000
TS15059-5000EA 263 USD InStock
TS15059-5000 TS15059-0250 TS15059-1000
L(-)-Malic acid 99%
L(-)-Malic acid


Formula: C₄H₆O₅
MW: 134.09 g/mol
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 99…107 °C
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00064213
CAS Number: 97-67-6
EINECS: 202-601-5
Order Now
Specification Test Results
Appearance White Crystalline powder or granules
Infrared spectrum Conforms
Melting point 99 to 107 °C
Sulfated ash ≤0.2 %
Specific optical rotation -1.5° to -3° (20 °C, 589 nm) (c=8.5, water)
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤300 ppm
Arsenic (As) ≤2 ppm
Heavy metals (as Pb) ≤20 ppm
Fumaric acid ≤0.2 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty