Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-α-Methylbenzylamine 99+%, (99% ee)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D(+)-α-Methylbenzylamine(R)-(+)-1-Phenylethylamine(R)-(+)-α-Phenylethylamine

15182-1000 15182-5000 15182-0250
TS15182-0250EA 70.2 USD InStock
TS15182-0250 TS15182-5000 TS15182-1000
(R)-(+)-α-Methylbenzylamine 99+%, (99% ee)
(R)-(+)-α-Methylbenzylamine


Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 184…186 °C (760 mmHg)
Melting Pt: –10 °C
Density: 0.95 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20°C, 589 nm)
GC ≥99.0 %
Enantiomeric excess ≥98.5 % (GC)
Specific optical rotation (20°C, 589 nm) > +37° (neat), (20°C, 589 nm) typically > +39°
Titration with HClO4 ≥98.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty