Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

15488-0500 15488-2500 15488-0100
TS15488-0500EA 144 USD InStock
TS15488-0500 TS15488-0100 TS15488-2500
Cyclopropylamine 99%
Cyclopropylamine


Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 49…50 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.824 g/cm³
Flash Pt: –25 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Volatile liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99 %
Titration with HCl 99.0 to 101.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty