Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Nitrophenyl)acetic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(2-Nitrophenyl)acetic acid

15706-1000 15706-0250
TS15706-0250EA 53 USD InStock
TS15706-0250 TS15706-1000
(2-Nitrophenyl)acetic acid 99%
(2-Nitrophenyl)acetic acid


Formula: (C₆H₄NO₂)CH₂COOH
MW: 181.15 g/mol
Melting Pt: 138…142 °C
MDL Number: MFCD00007190
CAS Number: 3740-52-1
EINECS: 223-128-0
Order Now
Specification Test Results
Appearance Yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 138 to 142 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty