Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Diaminocyclohexane cis- and trans-mixture 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1,2-Diaminocyclohexane1,2-Cyclohexanediamine

15810-0010 15810-2500
TS15810-2500EA 68 USD InStock
TS15810-2500 TS15810-0010
1,2-Diaminocyclohexane cis- and trans-mixture 99%
1,2-Diaminocyclohexane


Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 192 °C (1013 hPa)
Melting Pt: 2…15 °C
Density: 0.93 g/cm³
Flash Pt: 75 °C
MDL Number: MFCD00001491
CAS Number: 694-83-7
EINECS: 211-776-7
UN: 2735
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4920 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤75 APHA
Titration with HCl 99.4 to 100.6 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤150 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤2 ppm
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤50 ppm
Potassium (K) ≤50 ppm
Zinc (Zn) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty