Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tricine 98% for biochemistry

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acidTricine bufferN-(Tris(hydroxymethyl)methyl)glycine

17264-0250 17264-0010 17264-2500 17264-1000
TS17264-0250EA 35.49 USD InStock
TS17264-0250 TS17264-1000 TS17264-0010 TS17264-2500
Tricine 98% for biochemistry
Tricine


Formula: (HOCH₂)₃CNHCH₂CO₂H
MW: 179.17 g/mol
Melting Pt: 180…184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004277
CAS Number: 5704-04-1
EINECS: 227-193-6
Order Now
Specification Test Results
Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
UV (0.1 M in water)
UV at 260 nm A: ≤0.04
UV at 280 nm A: ≤0.02
pH 4.0 to 6.0 (1 % in water at 20°C)
Titration with HClO4 ≥97.5 % (On dried basis)
Heavy metals (as Pb) ≤5 ppm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty