Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Nitrophenyl)ethanol 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Nitrophenethyl alcoholp-Nitrophenethyl Alcohol

17961-0500 17961-0100
TS17961-0500EA 340 USD InStock
TS17961-0500 TS17961-0100
2-(4-Nitrophenyl)ethanol 99%
2-(4-Nitrophenyl)ethanol


Formula: C₈H₉NO₃
MW: 167.16 g/mol
Boiling Pt: 148…149 °C (2Torr)
Melting Pt: 56…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00010202
CAS Number: 100-27-6
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure
HPLC ≥97.0 %
Specific optical rotation +166° to +175° (20°C, 589 nm) (1 % in ethanol)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty