Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Diethylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

18106-0010 18106-2500 18106-0025
TS18106-0010EA 75 USD InStock
TS18106-0010 TS18106-2500 TS18106-0025
N,N-Diethylaniline 99%
N,N-Diethylaniline


Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 215.5 °C (1013 hPa)
Melting Pt: –38 °C
Density: 0.933 g/cm³
Flash Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00009042
CAS Number: 91-66-7
EINECS: 202-088-8
UN: 2432
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty