Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl isothiocyanatoformate 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ethoxycarbonyl isothiocyanate

18785-0250 18785-0050
TS18785-0250EA 236 USD InStock
TS18785-0250 TS18785-0050
Ethyl isothiocyanatoformate 95%
Ethyl isothiocyanatoformate


Formula: C₄H₅NO₂S
MW: 131.16 g/mol
Boiling Pt: 56 °C (18 mmHg)
Density: 1.16 g/cm³
Flash Pt: 50 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004814
CAS Number: 16182-04-0
UN: 2929
ADR: 6.1,II
Order Now
Specification Test Results
Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5020 (20°C, 589 nm)
GC ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty