Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Janus green B, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

19168-0250 19168-0050
TS19168-0050EA 78 USD InStock
TS19168-0050 TS19168-0250
Janus green B, pure, high purity biological stain
Janus green B


Formula: C₃₀H₃₁ClN₆
Storage Temperature: Ambient
MDL Number: MFCD00011758
CAS Number: 2869-83-2
Order Now
Specification Test Results
Appearance Dark green to dark brown to black Powder
Infrared spectrum Conforms
Dye content ≥50 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty