Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Aminonicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Aminopyridine-3-carboxylic acid6-Amino-3-pyridinecarboxylic acid

20254-0250 20254-0050 20254-0010
TS20254-0010EA 35 USD InStock
TS20254-0010 TS20254-0250 TS20254-0050
6-Aminonicotinic acid 98%
6-Aminonicotinic acid


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006326
CAS Number: 3167-49-5
EINECS: 221-630-4
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty