Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-2-chlorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

20323-0050
TS20323-0050EA 84 USD InStock
TS20323-0050
4-Amino-2-chlorobenzoic acid 97%
4-Amino-2-chlorobenzoic acid


Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 210…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5
Order Now
Specification Test Results
Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 210 to 215 °C
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty