Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Gulonic acid-γ-lactone 99%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Gulono γ-lactoneD-(-)-Gulono-1,4-lactoneD-Gulono-1,4-lactone(3R,4S,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one(3R,4S,5S)-5-((R)-1,2-Dihydroxyethyl)-3,4-dihydroxydihydrofuran-2(3H)-one

20445-0250
TS20445-0250EA 398 USD InStock
TS20445-0250
D-Gulonic acid-γ-lactone 99%
D-Gulonic acid-γ-lactone
Appearance: White Powder or crystals


Formula: C₆H₁₀O₆
MW: 178.14 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005391
CAS Number: 6322-07-2
Order Now
Specification Test Results
Appearance White Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥98.5 %
Specific optical rotation -52° to -58° (20°C, 589 nm) (c=4, water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty