Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-thyroxine 97+%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid

21566-0050 21566-0010 21566-2500
TS21566-0010EA 131 USD InStock
TS21566-0010 TS21566-0050 TS21566-2500
L-thyroxine 97+%
L-thyroxine


Formula: C₁₅H₁₁I₄NO₄
MW: 776.874 g/mol
Melting Pt: 235…236 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00002595
CAS Number: 51-48-9
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (110°C, 4 h)
HPLC ≥97.0 %
Specific optical rotation -4.0° to -6.0° (20°C, 589 nm) (c=1,1N NaOH:EtOH 1:2) (on dried substance)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty