Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Atropine sulfate Monohydrate 97+%

Supplier: Thermo Scientific Chemicals
Danger

22668-0100 22668-1000 22668-0250
TS22668-0100EA 139 USD InStock
TS22668-0100 TS22668-1000 TS22668-0250
Atropine sulfate Monohydrate 97+%
Atropine sulfate Monohydrate


Formula: C₁₇H₂₃NO₃
Melting Pt: >187 °C
Storage Temperature: Ambient
MDL Number: MFCD00074815
CAS Number: 5908-99-6
UN: 1544
ADR: 6.1,II
Order Now
Specification Test Results
Appearance White Powder or crystals
Infrared spectrum Conforms
Water 2.0 to 4.0 % (K.F.)
Residue after ignition ≤0.2 %
Specific optical rotation -0.50° to +0.05° (25°C, 589 nm) (c=10, water) (on anhydrous basis)
Titration with HClO4 97.0 to 103.0 % (On anhydrous basis)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty