Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclopropanemethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (Aminomethyl)cyclopropane(Cyclopropylmethyl)amineCyclopropylmethylamine

25872-0500 25872-0050 25872-0100
TS25872-0100EA 219 USD InStock
TS25872-0100 TS25872-0050 TS25872-0500
Cyclopropanemethylamine 97%
Cyclopropanemethylamine


Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 84…85 °C (760 mmHg)
Density: 0.82 g/cm³
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00037147
CAS Number: 2516-47-4
EINECS: 219-737-6
UN: 2733
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4300 to 1.4340 (20°C, 589 nm)
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty