Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gum arabic, powder

Supplier: Thermo Scientific Chemicals
Synonyms: Acacia gumAcacia

25885-0025 25885-2500 25885-0010
TS25885-2500EA 45.08 USD InStock
TS25885-2500 TS25885-0010 TS25885-0025
Gum arabic, powder
Gum arabic


Density: 1.35 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00081264
CAS Number: 9000-01-5
EINECS: 232-519-5
Order Now
Specification Test Results
Appearance White to yellow to light beige Powder
Infrared spectrum Conforms
Loss on drying ≤15.0 % (1 g, 105°C)
Additional info Agar, Starch, Tragacanth, dextrin, sucrose: absent
Identification Passes test (per Ph. Eur.)
Total ash ≤4.0 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤10 ppm
Heavy metals (as Pb) ≤40 ppm
Insoluble matter ≤0.1 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty