Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-4-hydroxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

27149-0050 27149-0250
TS27149-0250EA 120 USD InStock
TS27149-0250 TS27149-0050
3-Amino-4-hydroxybenzoic acid 98%
3-Amino-4-hydroxybenzoic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 198…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00007697
CAS Number: 1571-72-8
EINECS: 216-390-2
Order Now
Specification Test Results
Appearance Beige to grey to brown Crystalline powder
Infrared spectrum Conforms
Melting point 198 to 205 °C (Dec)
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty