Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Allylglycine 98%, 98% ee

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-2-Allylglycine(S)-(-)-alpha-AllylglycineH-Gly(Ally)-OH2-Allyl-L-glycine(2S)-2-Aminopent-4-enoic acid

27541-0010 27541-2500
TS27541-0010EA 278 USD InStock
TS27541-0010 TS27541-2500
L-Allylglycine 98%, 98% ee
L-Allylglycine


Formula: C₅H₉NO₂
MW: 115.132 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002627
CAS Number: 16338-48-0
Order Now
Specification Test Results
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
TLC ≥97.5 %
Enantiomeric excess ≥97.5 %
Specific optical rotation -35° to -39° (20°C, 589 nm) (c=4, water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty