Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromoanthranilic acid 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Amino-5-bromobenzoic acid

30562-0050 30562-0250
TS30562-0050EA 54 USD InStock
TS30562-0050 TS30562-0250
5-Bromoanthranilic acid 97%
5-Bromoanthranilic acid


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
Order Now
Specification Test Results
Appearance Beige Powder
Infrared spectrum Conforms
GC ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty