Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(diethylamino)phosphine 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (Diethylamino)phosphonous DichlorideDiethylphosphoramidous dichloride

31009-0050
TS31009-0050EA 72 USD InStock
TS31009-0050
Dichloro(diethylamino)phosphine 97%
Dichloro(diethylamino)phosphine
Appearance: Colorless to yellow Liquid


Formula: C₄H₁₀Cl₂NP
MW: 174.00 g/mol
Boiling Pt: 179 °C (760 mmHg)
Density: 1.196 g/cm³
Flash Pt: 82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013614
CAS Number: 1069-08-5
UN: 3265
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4960 to 1.4980 (20°C, 589 nm)
Titration Argentometric 96.0 to 104.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty