Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Amino-5-hydroxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

31031-0100 31031-0010
TS31031-0100EA 77 USD InStock
TS31031-0100 TS31031-0010
2-Amino-5-hydroxybenzoic acid 99%
2-Amino-5-hydroxybenzoic acid


Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347 °C (1013 hPa)
Melting Pt: 233…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00007870
CAS Number: 394-31-0
Order Now
Specification Test Results
Appearance Grey-brown Crystalline powder
Infrared spectrum Conforms
Titration with TBAH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty