Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methylumbelliferyl-β-D-glucopyranoside 99%

Supplier: Thermo Scientific Chemicals
Synonyms: 4-Methylumbelliferyl beta-D-GlucopyranosideMUGlucosid4-MU-β-D-Glc

33702-0025 33702-5000
TS33702-0025EA 704 USD InStock
TS33702-0025 TS33702-5000
4-Methylumbelliferyl-β-D-glucopyranoside 99%
4-Methylumbelliferyl-beta-D-glucopyranoside


Formula: C₁₆H₁₈O₈
MW: 338.312 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Freezer
MDL Number: MFCD00063694
CAS Number: 18997-57-4
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤2 % (K.F.)
TLC ≥98.5 %
Appearance of solution (3% soln. in acetone/water (2:1)) Clear, colorless
Specific optical rotation -97° to -101° (20°C, 589 nm) (c=0.3, water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty