Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-1,1'-Binaphthyl-2,2'-diamine(S)-(-)-2,2'-Diamino-1,1'-binaphthyl(S)-(-)-BINAM(S)-(-)-1,1'-Binaphthyl-2,2'-diamine(S)-(-)-1,1'-Bi(2-naphthylamine)

35652-0010
TS35652-0010EA 166 USD InStock
TS35652-0010
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene 97%
(S)-(-)-2,2'-Diamino-1,1'-binaphthalene


Formula: C₂₀H₁₆N₂
MW: 284.362 g/mol
Melting Pt: 242…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00145204
CAS Number: 18531-95-8
Order Now
Specification Test Results
Appearance White to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation -149.5° to -155.5° (20°C, 589 nm) (c=1, pyridine)
Titration with HClO4 96.0 to 104.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty