Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1R,2R)-(-)-1,2-Diaminocyclohexane 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (1R,2R)-Cyclohexane-1,2-diamine(R,R)-DACH

36464-0050 36464-0010
TS36464-0050EA 239.24 USD InStock
TS36464-0050 TS36464-0010
(1R,2R)-(-)-1,2-Diaminocyclohexane 99%
(1R,2R)-(-)-1,2-Diaminocyclohexane


Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 110…114 °C (40mbar)
Melting Pt: 40…45 °C
Flash Pt: 76 °C
Storage Temperature: Ambient
MDL Number: MFCD00062985
CAS Number: 20439-47-8
UN: 3259
ADR: 8,II
Order Now
Specification Test Results
Appearance White to light yellow Low melting crystalline solid
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥98.5 %
Specific optical rotation -24.5° to -26.5° (20 °C, 589 nm) (c=5, 1 N HCl)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty