Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Piperidin-3-ol 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Hydroxypiperidine3-Piperidinol

36708-0100 36708-0500
TS36708-0500EA 238 USD InStock
TS36708-0500 TS36708-0100
Piperidin-3-ol 98%
Piperidin-3-ol


Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 67…69 °C (2 mmHg)
Melting Pt: 56…62 °C
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014591
CAS Number: 6859-99-0
EINECS: 229-957-4
UN: 3263
ADR: 8,II
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder and/or lumps
Infrared spectrum Conforms
Melting point 56 to 62 °C
GC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty