Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(+)-α-Methylbenzylamine 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: D(+)-α-Methylbenzylamine(R)-(+)-1-Phenylethylamine(R)-(+)-α-Phenylethylamine

37146-0500 37146-0010 37146-2500
TS37146-0010EA 393 USD InStock
TS37146-0010 TS37146-2500 TS37146-0500
(R)-(+)-α-Methylbenzylamine 99+%
(R)-(+)-α-Methylbenzylamine
produced by BASF AG


Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 184…186 °C (760 mmHg)
Melting Pt: –10 °C
Density: 0.95 g/cm³
Flash Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD00064405
CAS Number: 3886-69-9
EINECS: 223-423-4
UN: 2922
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5270 (20°C, 589 nm)
Water ≤0.5 %
GC ≥99.0 %
Specific optical rotation +39° to +41° (20°C, 589 nm) (neat)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty