Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Benzylhydroxylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

39104-0010 39104-0050 39104-0250
TS39104-0010EA 35 USD InStock
TS39104-0010 TS39104-0250 TS39104-0050
N-Benzylhydroxylamine hydrochloride 98%
N-Benzylhydroxylamine hydrochloride


Formula: C₇H₁₀ClNO
MW: 159.6134 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00043377
CAS Number: 29601-98-7
Order Now
Specification Test Results
Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 108 to 112 °C
Titration Argentometric 97.5 to 102.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty