Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextran for biochemistry, low fraction

Supplier: Thermo Scientific Chemicals

40627-1000 40627-5000
TS40627-5000EA 479 USD InStock
TS40627-5000 TS40627-1000
Dextran for biochemistry, low fraction
Dextran


Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤7.0 %
UV (6 % in water)
UV at 375 nm A: ≤0.15
Viscosity 0.22 to 0.27 (1 % at 37°C)
Ash ≤2.0 %
Average molecular weight 60000 to 90000
pH 4.5 to 7.0 (6 % in water)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty