Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Histamine (free base) 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine

41171-0250 41171-0010 41171-0050
TS41171-0250EA 1144 USD InStock
TS41171-0250 TS41171-0050 TS41171-0010
Histamine (free base) 97%
Histamine (free base)


Formula: C₅H₉N₃
MW: 111.15 g/mol
Boiling Pt: 167 °C (.8 mmHg)
Melting Pt: 83…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00005210
CAS Number: 51-45-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 81 to 85 °C
HPLC ≥96.0 %
Titration with HClO4 ≥96.0 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty