Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Hydrazono-3-methylbenzothiazoline hydrochloride2-Hydrazono-3-methyl-2,3-dihydrobenzo[d]thiazole hydrochloride hydrate

41402-2500 41402-0500 41402-0100
TS41402-2500EA 1694 USD InStock
TS41402-2500 TS41402-0500 TS41402-0100
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate 98%
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
Appearance: White to beige Crystalline powder


Formula: C₈H₉N₃S·H₂O·HCl
MW: 233.714 g/mol
Melting Pt: 270…276 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149370
CAS Number: 38894-11-0
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 %
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty