Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Ethyl-5-nitroindole-2-carboxylate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Ethyl 5-nitro-1H-indole-2-carboxylate

42466-0025 42466-0100
TS42466-0025EA 125 USD InStock
TS42466-0025 TS42466-0100
Ethyl-5-nitroindole-2-carboxylate 95%
Ethyl-5-nitroindole-2-carboxylate


Formula: C₁₁H₁₀N₂O₄
MW: 234.211 g/mol
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00216477
CAS Number: 16732-57-3
Order Now
Specification Test Results
Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
Titration with TBAH ≥94 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty