Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethanolamine 99%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Monoethanolamine2-Aminoethanol

42725-1000 42725-8000
TS42725-1000EA 83.97 USD InStock
TS42725-1000 TS42725-8000
Ethanolamine 99%, AcroSeal™
Ethanolamine


Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 171 °C (1013 hPa)
Melting Pt: 10.3 °C
Density: 1.012 g/cm³
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
UN: 2491
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4530 to 1.4550 (20°C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤15 APHA

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty