Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethyltin chloride 25% (w/w) 1 M in hexanes, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorotrimethylstannane

42786-1000
TS42786-1000EA 187.96 USD InStock
TS42786-1000
Trimethyltin chloride 25% (w/w) 1 M in hexanes, AcroSeal™
Trimethyltin chloride


Formula: C₃H₉ClSn
MW: 199.27 g/mol
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD00000520
CAS Number: 1066-45-1
UN: 1992
ADR: 3,II
Order Now
Specification Test Results
Appearance Clear colorless Solution
Strength 0.90 to 1.10 M
Total trace metal impurities ≤50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty