Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyridoxamine dihydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride

43625-0010 43625-0050
TS43625-0010EA 77.98 USD InStock
TS43625-0010 TS43625-0050
Pyridoxamine dihydrochloride 98%
Pyridoxamine dihydrochloride


Formula: C₈H₁₂N₂O₂·2HCl
MW: 241.12 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Freezer
MDL Number: MFCD00012808
CAS Number: 524-36-7
Order Now
Specification Test Results
Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 % (on dry basis)
Water ≤10 % (K.F.)

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty