Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Dihydroorotic acid 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-2,6-Dioxohexahydropyrimidine-4-carboxylic acid

43724-0050 43724-0010
TS43724-0010EA 178 USD InStock
TS43724-0010 TS43724-0050
L-Dihydroorotic acid 99%
L-Dihydroorotic acid


Formula: C₅H₆N₂O₄
MW: 158.113 g/mol
Melting Pt: 254…255 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00085339
CAS Number: 5988-19-2
Order Now
Specification Test Results
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 253 to 257 °C
HPLC ≥98.5 %
Titration with NaOH ≥98.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty